Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-6-phosphate dehydrogenase, Bacillus sp. | 9001-40-5 | MFCD00081656 | 104 kDa | 10KU
SDP

Catalog No. 5000236584
Encompass_Preferred

Glucose-6-phosphate dehydrogenase from Bacillus sp. is a research-grade enzyme that catalyzes the oxidation of glucose-6-phosphate to 6-phosphogluconolactone with concurrent reduction of NADP+ to NADPH. Supplied as an activity-grade preparation (10 KU), it is intended for enzymatic assays, NADPH regeneration, and metabolic studies, including investigations of diabetes-related pathways.

  • Source organism: Bacillus sp.
  • Activity-grade pack: 10 KU
  • Catalyzes oxidation of glucose-6-phosphate, producing NADPH
  • Suitable for enzymatic assays and NADPH regeneration systems
  • Useful for metabolic and diabetes-related research
  • Provided as a research reagent; not for clinical use

Catalog No. 50-002-36584 Supplier Medchemexpress LLC Supplier No. HY125863C10KU
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose-6-phosphate dehydrogenase from Bacillus sp. is a research-grade enzyme that catalyzes the oxidation of glucose-6-phosphate to 6-phosphogluconolactone with concurrent reduction of NADP+ to NADPH. Supplied as an activity-grade preparation (10 KU), it is intended for enzymatic assays, NADPH regeneration, and metabolic studies, including investigations of diabetes-related pathways.

  • Source organism: Bacillus sp.
  • Activity-grade pack: 10 KU
  • Catalyzes oxidation of glucose-6-phosphate, producing NADPH
  • Suitable for enzymatic assays and NADPH regeneration systems
  • Useful for metabolic and diabetes-related research
  • Provided as a research reagent; not for clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.