Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose-6-phosphate dehydrogenase, Leuconostoc mesenteroides, Thermo Scientific Chemicals

Catalog No. AAJ601174I
Change view
Click to view available options
Quantity:
1 kU
10 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ601174I 1 kU
AAJ60117JPL 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ601174I Supplier Thermo Scientific Chemicals Supplier No. J601174I
Add to Cart

Glucose-6-phosphate dehydrogenase, derived from Leuconostoc mesenteroides, CAS# 9001-40-5, is an enzyme that catalyzes the conversion of p-glucose-6-phosphate to 6-phosphogluconolactone.

Glucose-6-phosphate dehydrogenase, Leuconostoc mesenteroides is used to convert p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+. It is used as an intermediate, chemical and research purpose and also used fo analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Glucose-6-phosphate dehydrogenase, derived from Leuconostoc mesenteroides, is a cytosolic enzyme used to convert p-glucose-6-phosphate to 6-phosphogluconolactone
  • This enzyme can use either NAD or NADP+ as an electron acceptor to promote this conversion

Application

  • Glucose-6-phosphate dehydrogenase belongs to the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells, including erythrocytes, by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH)
  • Glucose-6-phosphate dehydrogenase can be used to measure hexose in solution
  • It is used as an intermediate for chemical and research purposes
  • It is used as a model to test the effect of seed protein fractions on enzyme protection during dehydration
  • It has been used in assays for nicotinamide adenine dinucleotide and tissue pyridine nucleotides
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-40-5
MDL Number MFCD00081656
Synonym G6PD

Specifications

Packaging Plastic bottle
Quantity 1 kU
Solubility Information It is soluble in water and dilute buffer.
Physical Form Lyophilized Powder
Chemical Name or Material Glucose-6-phosphate dehydrogenase
EINECSNumber 232-602-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.