Learn More
Kuhner Shaker Inc GLUCOSE ARTICLE BEADS 25/PK
Catalog No. NC2743142
Encompass
FeedBeads, Glucose, 12 mm, 32 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. NC2743142 Supplier Kuhner Shaker Inc Supplier No. SMFB01001
FeedBeads, Glucose, 12 mm, 32 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!