Learn More
Crystal Chem Inc Glucose Assay Kit, 10-Kit Pack, 96-tests per well
SDP
Supplier:  Crystal Chem Inc 81697
Encompass_Preferred
The Glucose 10-kit pack is an economical option for high-volume users of our Glucose Assay Kit offered as ten individual kits.Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for human samples.
Catalog No. 50-194-7912