Learn More
Dojindo Molecular Technologies Inc Glucose Assay Kit-WST (200 TESTS) is used to determine intracellular levels of Glucose.
SDP
Catalog No. 501903720
Encompass_Preferred
Glucose, the most important biological energy source, is a known marker for energy metabolism. It is used not only as a marker for diabetes and obesity, but also for monitoring changes in intracellular metabolism such as lactate production. In addition, a recent study suggested that inhibition of the enzymatic activities involved in both glucose and lipid metabolism effectively inhibits cancer cell proliferation.Glucose Assay Kit-WST enables quantitation of glucose as a substrate in energy metabolism. This kit has been optimized to quantify glucose in cell culture supernatants by measuring absorbance in a colorimetric WST reaction. This kit contains a glucose standard solution that can be used to create a standard curve, it is formatted for 96-well microplates and the assay has a detection limit of 0.02 mmol/L glucose.
Catalog No. 50-190-3720 Supplier Dojindo Molecular Technologies Inc Supplier No. G26420
Glucose, the most important biological energy source, is a known marker for energy metabolism. It is used not only as a marker for diabetes and obesity, but also for monitoring changes in intracellular metabolism such as lactate production. In addition, a recent study suggested that inhibition of the enzymatic activities involved in both glucose and lipid metabolism effectively inhibits cancer cell proliferation.Glucose Assay Kit-WST enables quantitation of glucose as a substrate in energy metabolism. This kit has been optimized to quantify glucose in cell culture supernatants by measuring absorbance in a colorimetric WST reaction. This kit contains a glucose standard solution that can be used to create a standard curve, it is formatted for 96-well microplates and the assay has a detection limit of 0.02 mmol/L glucose.