Learn More
Sigma Aldrich Fine Chemicals Biosciences Glucose Assay Reagent, 20 mL
Catalog No. NC1704617
Encompass
For the quantitative determination of glucose in food and other materials utilizing the hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay.
Catalog No. NC1704617 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G329320ML
For the quantitative determination of glucose in food and other materials utilizing the hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay.