Learn More
Apexbio Technology LLC GLUCOSE GOD-POD KIT-48mg
SDP
Catalog No. 5000897342
Encompass_Preferred
Glucose Colorimetric Assay Kit (GOD-POD Method) is a widely used tool in biomedical research for quantifying glucose levels via a colorimetric detection method Originating from enzymatic analysis techniques this assay employs glucose oxidase (GOD) and peroxidase (POD) to catalyze glucose oxidation generating a measurable color change correlated with glucose concentration The mechanism of action involves the conversion of glucose into gluconic acid and hydrogen peroxide which reacts with a chromogenic substrate in the presence of POD This assay typically demonstrates activity in the nanomolar to low micromolar range and is employed in various applications including cell models animal studies and drug screening It provides a reliable method to assess glucose metabolism and evaluate potential therapeutic interventions Experimental concentrations typically depend on the specific research design ensuring adaptability across different experimental frameworks
Catalog No. 50-008-97342 Supplier Apexbio Technology LLC Supplier No. K230648
Glucose Colorimetric Assay Kit (GOD-POD Method) is a widely used tool in biomedical research for quantifying glucose levels via a colorimetric detection method Originating from enzymatic analysis techniques this assay employs glucose oxidase (GOD) and peroxidase (POD) to catalyze glucose oxidation generating a measurable color change correlated with glucose concentration The mechanism of action involves the conversion of glucose into gluconic acid and hydrogen peroxide which reacts with a chromogenic substrate in the presence of POD This assay typically demonstrates activity in the nanomolar to low micromolar range and is employed in various applications including cell models animal studies and drug screening It provides a reliable method to assess glucose metabolism and evaluate potential therapeutic interventions Experimental concentrations typically depend on the specific research design ensuring adaptability across different experimental frameworks