Learn More
Gibco™ Glucose, high-temperature short-time (HTST)-treated

Description
Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology. HTST is risk mitigation step that may reduce potential viral contaminations from entering upstream manufacturing from raw materials. Products such as glucose and other simple sugars are heated for a controlled period to not alter the treated product’s performance or viability. If HTST is a critical step in your process, outsourcing glucose can help you discover new efficiencies to reclaim time, space, and resources.
Product Features
- Multiple formulations and concentrations available
- Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
- Established products with set specifications are made to order
- Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
- Readily accessible documentation for accelerated qualification
Benefits
- Reduces time spent preparing buffers in-house
- Mitigates space requirement for equipment storage
- Streamlines in-house processes related to formulation, hydration, testing, and validation
- Meets workflow needs across stages of development and manufacturing of biologicals
Specifications
Specifications
| Color | Clear, colorless solution |
| Concentration | 40% (w/v) |
| Form | Liquid |
| Product Type | Glucose |
| Sterility | Sterile-filtered |
| Endotoxin Level | ≤1.00 |
| Format | Sample |
| Formulation | Glucose, HTST-treated, 40% (w/v) |
| Manufacturing Quality | cGMP |
| Quantity | 5L BPC |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.