Learn More
Description
- The Kit is designed for the quantitative measurement of glucose in serum, plasma, urine, or culture media.
- The enzymatic reaction provides high sensitivity and a broad linear range, making it suitable for metabolic research and high-throughput screening.
- Assay Technique: Photometry
- Hexokinase Activity: 1.0 unit/L
- Glucose-6-phosphate Dehydrogenase Activity: 1.0 unit/mL
- Glucose Standard Solution: 1.0 mg/mL concentration with 0.1% benzoic acid preservative
- Reconstitution: 0.02 L of water
- Capacity: 20 assays
- Applications: Metabolic research, disease modeling, and cannabis testing
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Product Type | Glucose Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.