Learn More
Creative Biomart GLUCOSE ISOMERASE, 1KG
Catalog No. NC2125392
Encompass
Glucose Isomerase from Streptomyces murinus, food grade: 5,000 U/g, 1KG
This item is not returnable. View return policy
Catalog No. NC2125392 Supplier Creative Biomart Supplier No. NATE0900
Glucose Isomerase from Streptomyces murinus, food grade: 5,000 U/g, 1KG