Learn More
Med Vet International Blood Glucose and Ketone Test Strips, Precision Xtra, 10 per Pack
Catalog No. 503043667
Encompass_Preferred
10-second test time and 1.5 microliters blood sample size End-fill or top-fill with visual confirmation Foil-wrapped test strips designed to prevent contamination errors due to exposure to air, moisture, or contamination 10 second test time and 1.5 microliters blood sample size
Catalog No. 50-304-3667 Supplier Med Vet International Supplier No. 647191BX
10-second test time and 1.5 microliters blood sample size End-fill or top-fill with visual confirmation Foil-wrapped test strips designed to prevent contamination errors due to exposure to air, moisture, or contamination 10 second test time and 1.5 microliters blood sample size