Learn More
QA Bio Inc Ludger Glucose Ladder - Procainamide
Catalog No. 502236612
Encompass
The procainamide glucose homopolymer (GHP) ladder derived from digested wheat starch used as a standard for sizing of glycans. Amount supplied sufficient for 30 HPLC runs. • The GHP ladder is also available labeled with 2-AB (CAB-GHP-30), 2-AA(CAA-GHP-30) and procainamide(PROC-GHP-30 . • Sources: Derived from wheat starch • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. • Amount: 100 picomoles • Purity: > 90% pure as assessed by a combination of HPLC
This item is not returnable. View return policy
Catalog No. 50-223-6612 Supplier QA Bio Inc Supplier No. CPROCGHP30
The procainamide glucose homopolymer (GHP) ladder derived from digested wheat starch used as a standard for sizing of glycans. Amount supplied sufficient for 30 HPLC runs. • The GHP ladder is also available labeled with 2-AB (CAB-GHP-30), 2-AA(CAA-GHP-30) and procainamide(PROC-GHP-30 . • Sources: Derived from wheat starch • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. • Amount: 100 picomoles • Purity: > 90% pure as assessed by a combination of HPLC