Learn More
ATHENA ES Glucose Nutrient Mix
Catalog No. 5000158101
Encompass_Preferred
Glucose Nutrient Mix is an APF Certified™ medium supplement used for enhancing the expression of proteins when using Athena’s Hyper Broth™ or Glucose M9Y. The powder blend consists of glucose as a carbon source and is Glucose Nutrient Mix Powder 150g 0109. Free Available in dry-powder packets. supplemented with calcium chloride and magnesium sulfate. Glucose Nutrient Mix is included with both media, but can now be purchased separately in individual packets.
Catalog No. 50-001-58101 Supplier ATHENA ES Supplier No. 0109
Glucose Nutrient Mix is an APF Certified™ medium supplement used for enhancing the expression of proteins when using Athena’s Hyper Broth™ or Glucose M9Y. The powder blend consists of glucose as a carbon source and is Glucose Nutrient Mix Powder 150g 0109. Free Available in dry-powder packets. supplemented with calcium chloride and magnesium sulfate. Glucose Nutrient Mix is included with both media, but can now be purchased separately in individual packets.