Learn More
Apexbio Technology LLC GLUCOSE O-TOLUIDINE KIT-1000mg
SDP
Catalog No. 5000897377
Encompass_Preferred
Glucose Colorimetric Assay Kit (O-toluidine Method) is utilized in the biochemical research category for glucose quantification commonly employed in clinical diagnostics and laboratory settings Originating from traditional methods of carbohydrate analysis this assay leverages the O-toluidine method to detect glucose concentration through the formation of a green chromogen The mechanism involves the reaction of glucose with O-toluidine under acidic conditions yielding a measurable colorimetric change This assay demonstrates in vitro activity within the nanomolar to low micromolar range making it suitable for high sensitivity glucose measurements It is typically applied in cell models animal studies and drug screening processes to assess glucose levels and metabolic function Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability for various experimental needs
Catalog No. 50-008-97377 Supplier Apexbio Technology LLC Supplier No. K23051000
Glucose Colorimetric Assay Kit (O-toluidine Method) is utilized in the biochemical research category for glucose quantification commonly employed in clinical diagnostics and laboratory settings Originating from traditional methods of carbohydrate analysis this assay leverages the O-toluidine method to detect glucose concentration through the formation of a green chromogen The mechanism involves the reaction of glucose with O-toluidine under acidic conditions yielding a measurable colorimetric change This assay demonstrates in vitro activity within the nanomolar to low micromolar range making it suitable for high sensitivity glucose measurements It is typically applied in cell models animal studies and drug screening processes to assess glucose levels and metabolic function Experimental concentrations typically depend on the specific research design ensuring flexibility and adaptability for various experimental needs