Learn More
NEOGEN Glucose OF Medium, 500 g
SDP
Catalog No. 502015591
Encompass_Preferred
Glucose OF Medium is used for the confirmation of Enterobacteriaceae in foods according to ISO 21528-1&2:2017, and is not intended for use in the diagnosis of disease or other conditions in humans.Enzymatic digest of casein provides nitrogen, vitamins, minerals and amino acids essential for growth. Sodium chloride supplies essential electrolytes for transport and osmotic balance. Dipotassium phosphate acts as a buffer system. Glucose is the fermentable carbohydrate providing carbon and energy. Bromothymol blue is the pH indicator.Colonies of presumptive Enterobacteriaceae should be confirmed by means of tests for the fermentation of glucose and the presence of oxidase. If the colonies are oxidase-negative and glucose-positive, the sample shall be regarded as being positive for Enterobacteriaceae.For more information visit https://www.neogen.com/solutions/Microbiology/glucose-of-medium
Catalog No. 50-201-5591 Supplier NEOGEN Supplier No. 700004539
Glucose OF Medium is used for the confirmation of Enterobacteriaceae in foods according to ISO 21528-1&2:2017, and is not intended for use in the diagnosis of disease or other conditions in humans.Enzymatic digest of casein provides nitrogen, vitamins, minerals and amino acids essential for growth. Sodium chloride supplies essential electrolytes for transport and osmotic balance. Dipotassium phosphate acts as a buffer system. Glucose is the fermentable carbohydrate providing carbon and energy. Bromothymol blue is the pH indicator.Colonies of presumptive Enterobacteriaceae should be confirmed by means of tests for the fermentation of glucose and the presence of oxidase. If the colonies are oxidase-negative and glucose-positive, the sample shall be regarded as being positive for Enterobacteriaceae.For more information visit https://www.neogen.com/solutions/Microbiology/glucose-of-medium