Learn More
TARGETMOL CHEMICALS INC Glucose oxidase 10MG
SDP
Catalog No. 502877995
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Glucose oxidase, an enzyme extracted from the fungus Aspergillus niger, is an important industrial enzyme in the food industry and is widely used in the deoxygenation of foods such as wine, beer, fruit juice, milk powder, and in the improvement of flour and prevention of browning of foods, etc. It has antioxidant and potential anticancer activities.
Catalog No. 50-287-7995 Supplier TARGETMOL CHEMICALS INC Supplier No. T7614810MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Glucose oxidase, an enzyme extracted from the fungus Aspergillus niger, is an important industrial enzyme in the food industry and is widely used in the deoxygenation of foods such as wine, beer, fruit juice, milk powder, and in the improvement of flour and prevention of browning of foods, etc. It has antioxidant and potential anticancer activities.