Learn More
Bioworld Glucose Oxidase, 100 KU
Supplier:  Bioworld 215600122
Encompass_Preferred
Glucose OxidaseMolecular Biology GradeSpecific activity (25° C): ≥95 U/mgSolubility (1%, water): Clear and haze-freeGlucose Oxidase: Catalase ratio >100:1Unit Definition: The amount of enzyme which causes the oxidation of 1 micromole of glucose per minute at 25 C, pH 7.0.50 kUActivity: 100,000 Units/g of solid
Catalog No. 50-198-975