Learn More
ALADDIN SCIENTIFIC CORPORATION Glucose Oxidase from Aspergill, 0.1MU
Catalog No. 5000082508
Encompass_Preferred
SPECIFICATIONS-Activity GO-2 100u/mg The activity may increase up to 160% if Ihe reaction mixture is saturated with oxygen Appearance Yellow powder lyophilizedContaminants Catalase 1u/mgStability Stable for 24 months at 5degCAdditives Sodium glutamateAPPLICATIONS -This enzyme is used for the enzymatic determination of glucosc in plasma by coupling with mutarotase and for amylase-activity assay by coupling with a-glucosidase (if mal tooligosaccharide or modified starch is used as a substrate) in clinical diagnosis PACKING-Polybottle in carton-casePRESERVATION-Below 5degC in a dry placc Fig 5
Catalog No. 50-000-82508 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. G1300840.1MU
SPECIFICATIONS-Activity GO-2 100u/mg The activity may increase up to 160% if Ihe reaction mixture is saturated with oxygen Appearance Yellow powder lyophilizedContaminants Catalase 1u/mgStability Stable for 24 months at 5degCAdditives Sodium glutamateAPPLICATIONS -This enzyme is used for the enzymatic determination of glucosc in plasma by coupling with mutarotase and for amylase-activity assay by coupling with a-glucosidase (if mal tooligosaccharide or modified starch is used as a substrate) in clinical diagnosis PACKING-Polybottle in carton-casePRESERVATION-Below 5degC in a dry placc Fig 5