Learn More
Abcam Glucose O x idase Conjugation Kit - Lightning-Link, 1MG
Catalog No. 502585602
Encompass_Preferred
Glucose Oxidase conjugation / labeling in < 4 hrs with 30 secs hands-on time using Glucose Oxidase Conjugation Kit - Lightning-Link ab102887. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5602 Supplier Abcam Supplier No. AB1028871MG
Glucose Oxidase conjugation / labeling in < 4 hrs with 30 secs hands-on time using Glucose Oxidase Conjugation Kit - Lightning-Link ab102887. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement