Learn More
Abcam Glucose O x idase Conjugation Kit - Lightning-Link, 3 x 10UG
Catalog No. 502585600
Encompass_Preferred
Glucose Oxidase conjugation / labeling in < 4 hrs with 30 secs hands-on time using Glucose Oxidase Conjugation Kit - Lightning-Link ab102887. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5600 Supplier Abcam Supplier No. AB1028873X10UG
Glucose Oxidase conjugation / labeling in < 4 hrs with 30 secs hands-on time using Glucose Oxidase Conjugation Kit - Lightning-Link ab102887. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement