Learn More
Crescent Chemical Co Inc GLUCOSE OXIDASE FROM A. NIGER
SDP
Supplier:  Crescent Chemical Co Inc SE22778.01
Encompass_Preferred
Glucose oxidase from Aspergillus niger min. 220 U/mg lyophil. Glucose oxidase is used in the assay of D-glucose (1). Ca. 300 U/mg protein. Size - 100MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 22778.01 CAS 9001-37-0
Catalog No. 50-247-639