Learn More
Crescent Chemical Co Inc GLUCOSE OXIDASE FROM A. NIGER
SDP
Catalog No. 50247640
Encompass_Preferred
Glucose oxidase from Aspergillus niger min. 220 U/mg lyophil. Glucose oxidase is used in the assay of D-glucose (1). Ca. 300 U/mg protein. Size - 500MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 22778.02 CAS 9001-37-0
Catalog No. 50-247-640 Supplier Crescent Chemical Co Inc Supplier No. SE22778.02
Glucose oxidase from Aspergillus niger min. 220 U/mg lyophil. Glucose oxidase is used in the assay of D-glucose (1). Ca. 300 U/mg protein. Size - 500MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 22778.02 CAS 9001-37-0