Learn More
Teco Diagnostics GLUCOSE OXIDASE TRINDER KIT
Catalog No. NC0820122
Encompass_Preferred
For the quantitative determination of total glucose in human serum. 4x250mL Vials of Glucose (Oxidase) 1x5mL Vial of Glucose Standard
Catalog No. NC0820122 Supplier Teco Diagnostics Supplier No. G5191L
For the quantitative determination of total glucose in human serum. 4x250mL Vials of Glucose (Oxidase) 1x5mL Vial of Glucose Standard