Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose-PEG2000-Cy3 | 95.0% | 10 MG
SDP

Catalog No. 5000298970
Encompass_Preferred

Glucose-PEG2000-Cy3 is a Cy3-labeled glucose-PEG conjugate designed for fluorescent labeling and delivery studies. The Cy3 fluorophore enables visible-range fluorescence detection, while the glucose-PEG2000 moiety promotes cellular uptake and can reduce endosomal degradation.

  • Cy3 fluorophore with emission maximum around 562-570 nm.
  • Peg2000 average molecular weight ~2000.
  • Purity typically 95.0% (by HPLC).
  • Suitable for immunolabeling, nucleic acid labeling, fluorescence microscopy, and flow cytometry.
  • Appearance: pale purple to purple oil.
  • Store protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-98970 Supplier Medchemexpress LLC Supplier No. HYD243310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose-PEG2000-Cy3 is a Cy3-labeled glucose-PEG conjugate designed for fluorescent labeling and delivery studies. The Cy3 fluorophore enables visible-range fluorescence detection, while the glucose-PEG2000 moiety promotes cellular uptake and can reduce endosomal degradation.

  • Cy3 fluorophore with emission maximum around 562-570 nm.
  • Peg2000 average molecular weight ~2000.
  • Purity typically 95.0% (by HPLC).
  • Suitable for immunolabeling, nucleic acid labeling, fluorescence microscopy, and flow cytometry.
  • Appearance: pale purple to purple oil.
  • Store protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.