Learn More
Bioworld GLUCOSE SEPAROPOREAGAROSE
Supplier:  Bioworld 201810762
Encompass
GLUCOSE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: -D-Glucose.; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow specificatio...
Catalog No. 50-197-865