Learn More
Medchemexpress LLC Glucose Transporter GLUT4 Antibody | 50 UL
SDP
Catalog No. 5000396010
Encompass_Preferred
Glucose Transporter GLUT4 Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to Glucose Transporter GLUT4.
  • Rabbit-derived and non-conjugated IgG polyclonal antibody
  • Targets Glucose Transporter GLUT4
  • Suitable for WB, IHC-P, and ELISA applications
  • Reacts with Human, Mouse, and Rat
Catalog No. 50-003-96010 Supplier Medchemexpress LLC Supplier No. HYP8068950UL
Glucose Transporter GLUT4 Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to Glucose Transporter GLUT4.
  • Rabbit-derived and non-conjugated IgG polyclonal antibody
  • Targets Glucose Transporter GLUT4
  • Suitable for WB, IHC-P, and ELISA applications
  • Reacts with Human, Mouse, and Rat