Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose Transporter GLUT4 Antibody | 55 kDa | 10 UL
SDP

Catalog No. 5000395764
Encompass_Preferred

Glucose Transporter GLUT4 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody targeting Glucose Transporter GLUT4. This product is for research use only and is supplied as a liquid.

  • Applications include WB, IHC-P, and ELISA.
  • Reactivity with human, mouse, and rat samples.
  • Stable under recommended storage conditions.
  • Does not contain hazardous substances or mixtures.
  • Target protein band size: 55 kDa.

Catalog No. 50-003-95764 Supplier Medchemexpress LLC Supplier No. HYP8068910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose Transporter GLUT4 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody targeting Glucose Transporter GLUT4. This product is for research use only and is supplied as a liquid.

  • Applications include WB, IHC-P, and ELISA.
  • Reactivity with human, mouse, and rat samples.
  • Stable under recommended storage conditions.
  • Does not contain hazardous substances or mixtures.
  • Target protein band size: 55 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.