Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucose Transporter GLUT4 Antibody (YA753) | Predicted band size: 55 kDa; Observed band size: 55 kDa | 100 UL
SDP

Catalog No. 5000396253
Encompass_Preferred

Glucose Transporter GLUT4 Antibody (YA753) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Glucose Transporter GLUT4. This antibody is designed for research use only and plays a crucial role in removing glucose from circulation, regulated by insulin.

  • Host is mouse
  • Clonality is monoclonal IgG1
  • Targets Glucose Transporter GLUT4
  • Reactivity with human and mouse
  • Immunogen affinity purified
  • Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • Applications include Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunohistochemistry-Paraffin (IHC-P)

Catalog No. 50-003-96253 Supplier Medchemexpress LLC Supplier No. HYP80495100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucose Transporter GLUT4 Antibody (YA753) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Glucose Transporter GLUT4. This antibody is designed for research use only and plays a crucial role in removing glucose from circulation, regulated by insulin.

  • Host is mouse
  • Clonality is monoclonal IgG1
  • Targets Glucose Transporter GLUT4
  • Reactivity with human and mouse
  • Immunogen affinity purified
  • Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • Applications include Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunohistochemistry-Paraffin (IHC-P)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.