Learn More
Dojindo Molecular Technologies Inc Glucose Uptake Assay Kit-Blue (1 set). Highly sensitive and simple measurement of glucose uptake capacity. No more radioactive waste. High cellular retention. Ex=386 nm, Em=474 nm.
SDP
Catalog No. NC2004940
Encompass_Preferred
Glucose uptake capacity is an important indicator in the field of diabetes research. Diabetes is caused by decreased insulin secretion and responsiveness, and it inhibits cellular uptake of glucose. One common method for evaluating the glucose uptake ability of cells uses radioisotope-labeled glucose. Although this method has been used for many years, it requires special handling facilities and disposal of radioactive materials, at the disadvantage of inconvenience and high cost. Glucose Uptake Probe-Blue has resolved these limitations. This probe emits strong blue fluorescence (Ex=386 nm, Em=474 nm), allowing for easy detection of glucose uptake by fluorescent microscope and flow cytometry. It is not only a boon in regards to potential multiplex capabilities, but it also avoids emission overlap with the autofluorescence that cells produce. Dojindo offers various glucose uptake assay kits. (https://dojindo.com/glucose-uptake-assay-kit-blue-green-red/)
Catalog No. NC2004940 Supplier Dojindo Molecular Technologies Inc Supplier No. UP0110
Add to Cart
Add to Cart
Glucose uptake capacity is an important indicator in the field of diabetes research. Diabetes is caused by decreased insulin secretion and responsiveness, and it inhibits cellular uptake of glucose. One common method for evaluating the glucose uptake ability of cells uses radioisotope-labeled glucose. Although this method has been used for many years, it requires special handling facilities and disposal of radioactive materials, at the disadvantage of inconvenience and high cost. Glucose Uptake Probe-Blue has resolved these limitations. This probe emits strong blue fluorescence (Ex=386 nm, Em=474 nm), allowing for easy detection of glucose uptake by fluorescent microscope and flow cytometry. It is not only a boon in regards to potential multiplex capabilities, but it also avoids emission overlap with the autofluorescence that cells produce. Dojindo offers various glucose uptake assay kits. (https://dojindo.com/glucose-uptake-assay-kit-blue-green-red/)