Learn More
Dojindo Molecular Technologies Inc Glucose Uptake Assay Kit-Red (1 set). Highly sensitive and simple measurement of glucose uptake capacity. No more radioactive waste. High cellular retention. Ex=560 nm, Em=572 nm.
SDP
Catalog No. NC2004942
Encompass_Preferred
Glucose uptake capacity is an important indicator in the field of diabetes research. Diabetes is caused by decreased insulin secretion and responsiveness, and it inhibits cellular uptake of glucose. One common method for evaluating the glucose uptake ability of cells uses radioisotope-labeled glucose. Although this method has been used for many years, it requires special handling facilities and disposal of radioactive materials, at the disadvantage of inconvenience and high cost. Glucose Uptake Probe-Red has resolved these limitations. This probe emits strong red fluorescence (Ex=560 nm, Em=572 nm), allowing for easy detection of glucose uptake by a fluorescent microscope, microplate reader, and flow cytometry. It is not only a boon in regards to potential multiplex capabilities, but it also avoids emission overlap with the autofluorescence that cells produce. Dojindo offers various glucose uptake assay kits. (https://dojindo.com/glucose-uptake-assay-kit-blue-green-red/)
Catalog No. NC2004942 Supplier Dojindo Molecular Technologies Inc Supplier No. UP0310
Glucose uptake capacity is an important indicator in the field of diabetes research. Diabetes is caused by decreased insulin secretion and responsiveness, and it inhibits cellular uptake of glucose. One common method for evaluating the glucose uptake ability of cells uses radioisotope-labeled glucose. Although this method has been used for many years, it requires special handling facilities and disposal of radioactive materials, at the disadvantage of inconvenience and high cost. Glucose Uptake Probe-Red has resolved these limitations. This probe emits strong red fluorescence (Ex=560 nm, Em=572 nm), allowing for easy detection of glucose uptake by a fluorescent microscope, microplate reader, and flow cytometry. It is not only a boon in regards to potential multiplex capabilities, but it also avoids emission overlap with the autofluorescence that cells produce. Dojindo offers various glucose uptake assay kits. (https://dojindo.com/glucose-uptake-assay-kit-blue-green-red/)