Learn More
Cayman Chemical Glucose Uptake CelBased As 1ea
SDP
Catalog No. 502897019
Encompass_Preferred
Cayman’s Glucose Uptake Cell-Based Assay Kit provides a convenient tool for studying modulators of cellular glucose uptake. The kit employs NBD-glucose (2-NBDG), a fluorescently labeled deoxyglucose analog, as a probe for the detection of glucose taken up by cultured cells. Researchers interested in cellular metabolism, such as cancer biologists, immunologists, physiologists, and others, will find this kit a robust tool for measuring energy consumption in living cells.
Catalog No. 50-289-7019 Supplier Cayman Chemical Supplier No. 6004701 EA
Cayman’s Glucose Uptake Cell-Based Assay Kit provides a convenient tool for studying modulators of cellular glucose uptake. The kit employs NBD-glucose (2-NBDG), a fluorescently labeled deoxyglucose analog, as a probe for the detection of glucose taken up by cultured cells. Researchers interested in cellular metabolism, such as cancer biologists, immunologists, physiologists, and others, will find this kit a robust tool for measuring energy consumption in living cells.