Learn More
HIMEDIA GLUCOSE YEAST EXTRACT BC AGAR
Catalog No. NC2292276
Encompass_Preferred
Glucose Yeast extract BC Agar Medium 500 grams.Glucose Yeast Extract BC Agar Medium is recommended for enumeration and cultivation of Bacillus coagulans from food samples. It is also used in routine diagnosis for spoilage in canned foods. The unique property of Bacillus coagulans of forming spores can be identified on this medium on incubation at higher temperature as 40 ± 2°C for 48 hours.
This item is not returnable. View return policy
Catalog No. NC2292276 Supplier HIMEDIA Supplier No. M2102500G
Glucose Yeast extract BC Agar Medium 500 grams.Glucose Yeast Extract BC Agar Medium is recommended for enumeration and cultivation of Bacillus coagulans from food samples. It is also used in routine diagnosis for spoilage in canned foods. The unique property of Bacillus coagulans of forming spores can be identified on this medium on incubation at higher temperature as 40 ± 2°C for 48 hours.