Learn More
Medchemexpress LLC Glucosylceramide | 85305-87-9 | 98% | 5 MG
SDP
Catalog No. 5000391641
Encompass_Preferred
Glucosylceramides are sphingolipid molecules found in both neuronal and non-neuronal mammalian tissues, as well as in low quantities across various plant species. Elevated levels of glucosylceramides provide cellular protection and prepare certain cells for proliferation; however, they have also been linked to obesity-induced insulin resistance in mice and to neuronal deficits observed in neuronopathic Gaucher disease.
  • Found in neuronal and non-neuronal mammalian tissues.
  • Also present in low quantities across various plant species.
  • Elevated levels provide cellular protection.
  • Prepares certain cells for proliferation.
Catalog No. 50-003-91641 Supplier Medchemexpress LLC Supplier No. HY1650355MG
Glucosylceramides are sphingolipid molecules found in both neuronal and non-neuronal mammalian tissues, as well as in low quantities across various plant species. Elevated levels of glucosylceramides provide cellular protection and prepare certain cells for proliferation; however, they have also been linked to obesity-induced insulin resistance in mice and to neuronal deficits observed in neuronopathic Gaucher disease.
  • Found in neuronal and non-neuronal mammalian tissues.
  • Also present in low quantities across various plant species.
  • Elevated levels provide cellular protection.
  • Prepares certain cells for proliferation.