Learn More
Medchemexpress LLC Glucosylceramide synthase-IN-3 | 3029827-91-3 | 99.11% | C21H20FN3O3 | 1 ML
SDP
Catalog No. 5000354838
Encompass_Preferred
Glucosylceramide synthase-IN-3 (compound BZ1) is a potent, brain-penetrant, and orally active glucosylceramide synthase (GCS) inhibitor with an IC50 of 16 nM for human GCS. It can be used for Gaucher's disease research.
  • Storage conditions for powder: -20°C for 3 years
  • Storage conditions for powder: 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months
  • Storage conditions in solvent: -20°C for 1 month
Catalog No. 50-003-54838 Supplier Medchemexpress LLC Supplier No. HY14427010MM/1ML
Glucosylceramide synthase-IN-3 (compound BZ1) is a potent, brain-penetrant, and orally active glucosylceramide synthase (GCS) inhibitor with an IC50 of 16 nM for human GCS. It can be used for Gaucher's disease research.
  • Storage conditions for powder: -20°C for 3 years
  • Storage conditions for powder: 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months
  • Storage conditions in solvent: -20°C for 1 month