Learn More
Cayman Chemical Glucosylceramide (bovine buttermilk), Purity: ≥98%, 50 mg
SDP
Catalog No. 502993783
Encompass_Preferred
A mixture of glucosylceramides isolated from soy; levels decrease during cold acclimatization in plants; involved in the regulation of virulence in fungi and act as elicitors in plants, stimulating plant defense mechanisms; increased levels are associated with obesity-induced insulin resistance in mice and with neuronal deficits observed in neuronopathic Gaucher disease
Catalog No. 50-299-3783 Supplier Cayman Chemical Supplier No. 2320650 MG
A mixture of glucosylceramides isolated from soy; levels decrease during cold acclimatization in plants; involved in the regulation of virulence in fungi and act as elicitors in plants, stimulating plant defense mechanisms; increased levels are associated with obesity-induced insulin resistance in mice and with neuronal deficits observed in neuronopathic Gaucher disease