Learn More
Cayman Chemical Glucosylceramide (plant), Purity: ≥98%, CAS: 497155-61-0, 100 mg
SDP
Catalog No. 502934958
Encompass_Preferred
A mixture of glucosylceramides isolated from soy; levels decrease during cold acclimatization in plants; involved in the regulation of virulence in fungi and act as elicitors in plants, stimulating plant defense mechanisms; increased levels are associated with obesity-induced insulin resistance in mice and with neuronal deficits observed in neuronopathic Gaucher disease
Catalog No. 50-293-4958 Supplier Cayman Chemical Supplier No. 25850100 MG
A mixture of glucosylceramides isolated from soy; levels decrease during cold acclimatization in plants; involved in the regulation of virulence in fungi and act as elicitors in plants, stimulating plant defense mechanisms; increased levels are associated with obesity-induced insulin resistance in mice and with neuronal deficits observed in neuronopathic Gaucher disease