Learn More
Cayman Chemical Glucotropaeolin (potassium salt), Purity: ≥98%, CAS: 5115-71-9, 10 mg
SDP
Catalog No. 502964447
Encompass_Preferred
A glucosinolate that has been found in cruciferous plants such as L. sativum; reduces the number of HM1-IMSS E. histolytica trophozoites (IC50 = 20.4 µg/ml)
Catalog No. 50-296-4447 Supplier Cayman Chemical Supplier No. 2146310 MG
A glucosinolate that has been found in cruciferous plants such as L. sativum; reduces the number of HM1-IMSS E. histolytica trophozoites (IC50 = 20.4 µg/ml)