Learn More
Medchemexpress LLC Glucuronic acid sodium | 7182-77-6 | 1 MG
SDP
Catalog No. 5000420093
Encompass_Preferred
Glucuronic acid sodium is a polysaccharide that exhibits anti-inflammatory activity, supporting the healing of ulcerative colitis. It also enhances the intestinal barrier by upregulating tight junction proteins like ZO-1 and Occludin. Additionally, it modulates gut microbiota by increasing beneficial bacterial populations while suppressing harmful ones.
  • Exhibits anti-inflammatory activity
  • Supports healing of ulcerative colitis
  • Enhances intestinal barrier function
  • Modulates gut microbiota
  • Increases beneficial bacterial populations
  • Suppresses harmful bacterial populations
Catalog No. 50-004-20093 Supplier Medchemexpress LLC Supplier No. HYN6612B1MG
Glucuronic acid sodium is a polysaccharide that exhibits anti-inflammatory activity, supporting the healing of ulcerative colitis. It also enhances the intestinal barrier by upregulating tight junction proteins like ZO-1 and Occludin. Additionally, it modulates gut microbiota by increasing beneficial bacterial populations while suppressing harmful ones.
  • Exhibits anti-inflammatory activity
  • Supports healing of ulcerative colitis
  • Enhances intestinal barrier function
  • Modulates gut microbiota
  • Increases beneficial bacterial populations
  • Suppresses harmful bacterial populations