Learn More
MOLECULAR DEPOT LLC Glucuronidase-ArylSulf. 5 mL
Catalog No. 5000013374
Encompass_Preferred
b-Glucuronidase/Arylsulfatase from Helix pomatia is an enzymatic bend of -Glucuronidase and Arylsulfatase from Helix pomatia with each enzyme having an activity of at least 3500 U/mL This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13374 Supplier MOLECULAR DEPOT LLC Supplier No. B2014879
b-Glucuronidase/Arylsulfatase from Helix pomatia is an enzymatic bend of -Glucuronidase and Arylsulfatase from Helix pomatia with each enzyme having an activity of at least 3500 U/mL This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only