Learn More
Cayman Chemical β-Glucuronidase Inhibitor, Purity: ≥98%, CAS: 484006-66-8, 1 mg
SDP
Catalog No. 502944868
Encompass_Preferred
An inhibitor of bacterial β-glucuronidase (IC50 = 283 nM for the E. coli enzyme); inhibits wild-type but not Δ360-376 E. coli β-glucuronidase at 100 µM; reduces irinotecan-induced colonic epithelial damage and the number of bloody diarrheal events in a mouse model of chemotherapy-induced intestinal toxicity at 50 mg/kg
Catalog No. 50-294-4868 Supplier Cayman Chemical Supplier No. 412281 MG
An inhibitor of bacterial β-glucuronidase (IC50 = 283 nM for the E. coli enzyme); inhibits wild-type but not Δ360-376 E. coli β-glucuronidase at 100 µM; reduces irinotecan-induced colonic epithelial damage and the number of bloody diarrheal events in a mouse model of chemotherapy-induced intestinal toxicity at 50 mg/kg