Learn More
Cayman Chemical γ-Glu-Cys (ammonium salt), Purity: ≥95%, 100 mg
SDP
Catalog No. 502991276
Encompass_Preferred
An intermediate in GSH synthesis; formed by ATP-dependent condensation of cysteine and glutamate by γ-glutamylcysteine synthetase; an essential component of phytochelatins
Catalog No. 50-299-1276 Supplier Cayman Chemical Supplier No. 30569100 MG
An intermediate in GSH synthesis; formed by ATP-dependent condensation of cysteine and glutamate by γ-glutamylcysteine synthetase; an essential component of phytochelatins