Learn More
Cayman Chemical GluCys ammonium salt 10mg
SDP
Catalog No. 502992193
Encompass_Preferred
An intermediate in GSH synthesis; formed by ATP-dependent condensation of cysteine and glutamate by γ-glutamylcysteine synthetase; an essential component of phytochelatins
Catalog No. 50-299-2193 Supplier Cayman Chemical Supplier No. 3056910 MG
An intermediate in GSH synthesis; formed by ATP-dependent condensation of cysteine and glutamate by γ-glutamylcysteine synthetase; an essential component of phytochelatins