Learn More
Aviva Systems Biology Corp GLUD1 Antibody - N-terminal region (ARP45708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLUD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLUD1. Alternate names: GDH, GDH1, GLUD, MGC132003ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-59110