Learn More
Aviva Systems Biology Corp GLUD2 Antibody - N-terminal region (ARP54828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLUD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLUD2. Alternate names: GDH2, GLUDP1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-62897