Learn More
Chem Service Inc Glufosinate-ammonium,77182-82-2,100MG
SDP
Catalog No. NC1044687
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula C5H15N2O4P, Purity 98.20%, Molecular Weight 198.2, Melting Point 215 C, Snynonyms: D,L-Phosphinothricin
Catalog No. NC1044687 Supplier Chem Service Inc Supplier No. N12111100MG
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula C5H15N2O4P, Purity 98.20%, Molecular Weight 198.2, Melting Point 215 C, Snynonyms: D,L-Phosphinothricin