Learn More
Medchemexpress LLC Glufosinate-d3 hydrochloride | 1323254-05-2 | C5H10D3ClNO4P | 1 MG
SDP
Catalog No. 5000373665
Encompass_Preferred
Glufosinate-d3 hydrochloride is a deuterium labeled compound. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for use as tracers in the drug development process. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through various methods such as NMR, GC-MS, or LC-MS.
Catalog No. 50-003-73665 Supplier Medchemexpress LLC Supplier No. HY152055S1MG
Glufosinate-d3 hydrochloride is a deuterium labeled compound. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for use as tracers in the drug development process. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through various methods such as NMR, GC-MS, or LC-MS.