Learn More
Cayman Chemical Glufosinate (ammonium salt), Purity: ≥95%, CAS: 77182-82-2, 100 mg
SDP
Catalog No. 502965705
Encompass_Preferred
A broad-spectrum herbicide; an irreversible inhibitor of GS1 and GS2 (Kis = 1.1. and 4.8 µM, respectively); reduces dry biomass of the weeds E. canadensis and L. rigidum (ED50s = 26 and 763 g/hectare, respectively); has no observed adverse effects in rats and mice at 4.1 and 17 mg/kg per day for three months
Catalog No. 50-296-5705 Supplier Cayman Chemical Supplier No. 16675100 MG
A broad-spectrum herbicide; an irreversible inhibitor of GS1 and GS2 (Kis = 1.1. and 4.8 µM, respectively); reduces dry biomass of the weeds E. canadensis and L. rigidum (ED50s = 26 and 763 g/hectare, respectively); has no observed adverse effects in rats and mice at 4.1 and 17 mg/kg per day for three months