Learn More
Sino Biological Anti-Glutamine Synthetase/GLUL Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107911T0820
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Cairo spiny mouse Glutamine Synthetase, and purified by antigen affinity chromatography.
Catalog No. 50-241-1561