Learn More
Medchemexpress LLC HY-116000 10mg Medchemexpress, Glumetinib CAS:1642581-63-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11600010MG
Encompass_Preferred
Medchemexpress, HY-116000 10mg Glumetinib CAS:1642581-63-2 Glumetinib (SCC244) is a potent and highly selective c-Met kinase inhibitor with an IC50 of 0.42 nM. Glumetinib shows antitumor activity and a superior safety margin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9400