Learn More
Medchemexpress LLC GluN1(356-385) | 99.95% | 3415.90 | 5 MG
SDP
Catalog No. 5000396327
Encompass_Preferred
GluN1 (356-385) is an antigenic peptide used against N-methyl-D-aspartate receptor (NMDAR) encephalitis. It effectively reduces the density of surface NMDAR clusters in hippocampal neurons. This peptide can be utilized to investigate the pathogenesis of anti-NMDAR encephalitis.
  • Antigenic peptide against N-methyl-D-aspartate receptor (NMDAR) encephalitis.
  • Reduces the density of surface NMDAR clusters in hippocampal neurons.
  • Useful for studying the pathogenesis of anti-NMDAR encephalitis.
  • Purity of 99.95%.
Catalog No. 50-003-96327 Supplier Medchemexpress LLC Supplier No. HYP59125MG
GluN1 (356-385) is an antigenic peptide used against N-methyl-D-aspartate receptor (NMDAR) encephalitis. It effectively reduces the density of surface NMDAR clusters in hippocampal neurons. This peptide can be utilized to investigate the pathogenesis of anti-NMDAR encephalitis.
  • Antigenic peptide against N-methyl-D-aspartate receptor (NMDAR) encephalitis.
  • Reduces the density of surface NMDAR clusters in hippocampal neurons.
  • Useful for studying the pathogenesis of anti-NMDAR encephalitis.
  • Purity of 99.95%.